ENA429359782 Enamine

ethyl({[1-(prop-2-yn-1-yl)-1H-pyrazol-3-yl]methyl})amine hydrochloride

  • MDL number MFCD28118741
  • Cas Number : 1803586-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188648
Availability details for
Quantity
  • Availability
  • Add to Cart