ENA429359785 Enamine

5-(5-bromothiophen-2-yl)-1,2,4-triazin-3-amine

  • MDL number MFCD12819877
  • Cas Number : 1183278-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188651
Availability details for
Quantity
  • Availability
  • Add to Cart