ENA429359789 Enamine

4-amino-1-ethyl-N-methyl-N-[(2-methyl-1,3-thiazol-4-yl)methyl]-1H-pyrazole-5-carboxamide

  • MDL number MFCD20796733
  • Cas Number : 1485265-60-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188657

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188657
Availability details for
Quantity
  • Availability
  • Add to Cart