ENA429359790 Enamine

5-phenyl-2,3-dihydro-1,2,4-oxadiazol-3-one

  • MDL number MFCD00956164
  • Cas Number : 21084-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188658

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188658
Availability details for
Quantity
  • Availability
  • Add to Cart