ENA429359791 Enamine

5-benzyl-1,2,4-oxadiazol-3-amine

  • MDL number MFCD00207921
  • Cas Number : 35604-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188659

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188659
Availability details for
Quantity
  • Availability
  • Add to Cart