ENA429359793 Enamine

3-(1-benzothiophen-2-yl)-3-oxopropanenitrile

  • MDL number MFCD14660419
  • Cas Number : 1249556-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188661

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188661
Availability details for
Quantity
  • Availability
  • Add to Cart