ENA429359797 Enamine

2-bromo-1-cycloheptylethan-1-one

  • MDL number MFCD13173386
  • Cas Number : 155412-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188666

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188666
Availability details for
Quantity
  • Availability
  • Add to Cart