ENA429359800 Enamine

4-(5-amino-1-methyl-1H-1,2,4-triazol-3-yl)pyridin-2-amine

  • MDL number MFCD26917443
  • Cas Number : 1566546-85-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188669

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188669
Availability details for
Quantity
  • Availability
  • Add to Cart