ENA429359807 Enamine

3-(dimethylamino)-1-(4-fluoro-2-hydroxyphenyl)prop-2-en-1-one

  • Cas Number : 138469-95-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188698

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188698
Availability details for
Quantity
  • Availability
  • Add to Cart