ENA429359813 Enamine

(2,6-dichloro-4-fluorophenyl)boronic acid

  • MDL number MFCD20526365
  • Cas Number : 1451392-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188705

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-188705
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart