ENA429359814 Enamine

1-(3-amino-2-methylphenyl)-3-ethylurea

  • MDL number MFCD11213431
  • Cas Number : 1094663-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188706

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188706
Availability details for
Quantity
  • Availability
  • Add to Cart