ENA429359816 Enamine

methyl 2-chloro-3-(phenylformamido)propanoate

  • MDL number MFCD28118747
  • Cas Number : 1702112-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188709
Availability details for
Quantity
  • Availability
  • Add to Cart