ENA429359823 Enamine

3-methyl-1-(naphthalene-1-carbonyl)-1H-1,2,4-triazol-5-amine

  • MDL number MFCD28118750
  • Cas Number : 1333724-04-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188720
Availability details for
Quantity
  • Availability
  • Add to Cart