ENA429359825 Enamine

2-(2,2-difluorocyclopentyl)acetic acid

  • MDL number MFCD28118751
  • Cas Number : 1532703-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188724

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188724
Availability details for
Quantity
  • Availability
  • Add to Cart