ENA429359835 Enamine

1-(4-hydroxy-2-methylphenyl)propan-1-one

  • MDL number MFCD19309257
  • Cas Number : 2887-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188737

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188737
Availability details for
Quantity
  • Availability
  • Add to Cart