ENA429359836 Enamine

2-bromo-4-(trifluoromethyl)pyridin-3-ol

  • MDL number MFCD16610032
  • Cas Number : 1211526-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188738

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188738
Availability details for
Quantity
  • Availability
  • Add to Cart