ENA429359839 Enamine

ethyl 7-oxoazepane-2-carboxylate

  • MDL number MFCD24549430
  • Cas Number : 38167-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188741

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-188741
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart