ENA429359844 Enamine

3-(3-iodo-1H-1,2,4-triazol-1-yl)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD23768906
  • Cas Number : 1520131-57-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188750
Availability details for
Quantity
  • Availability
  • Add to Cart