ENA429359845 Enamine

1-[4-(trifluoromethyl)cyclohexyl]ethan-1-one

  • MDL number MFCD21775950
  • Cas Number : 916481-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188751
Availability details for
Quantity
  • Availability
  • Add to Cart