ENA429359847 Enamine

1-(5-bromopyridin-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD21194072
  • Cas Number : 1494496-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188753

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188753
Availability details for
Quantity
  • Availability
  • Add to Cart