ENA429359850 Enamine

3-(2-fluorophenyl)-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD28118757
  • Cas Number : 1788613-48-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188757

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188757
Availability details for
Quantity
  • Availability
  • Add to Cart