ENA429359851 Enamine

methyl 2-amino-2-(1-phenyl-1H-pyrazol-4-yl)acetate hydrochloride

  • MDL number MFCD28118758
  • Cas Number : 1803562-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188770
Availability details for
Quantity
  • Availability
  • Add to Cart