ENA429359860 Enamine

methyl 7-fluoro-4-methoxy-1H-indole-2-carboxylate

  • MDL number MFCD18251688
  • Cas Number : 1803588-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188795

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188795
Availability details for
Quantity
  • Availability
  • Add to Cart