ENA429359862 Enamine

6-(5-fluoropyridin-3-yl)-N-methylpyridazin-3-amine

  • MDL number MFCD21277804
  • Cas Number : 1489634-55-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188827

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188827
Availability details for
Quantity
  • Availability
  • Add to Cart