ENA429359866 Enamine

N-(3-amino-4-methoxyphenyl)prop-2-ynamide

  • MDL number MFCD12084005
  • Cas Number : 1153544-51-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188851

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188851
Availability details for
Quantity
  • Availability
  • Add to Cart