ENA429359867 Enamine

2-chloro-7-fluoroquinoxaline

  • MDL number MFCD22571052
  • Cas Number : 1233932-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188852

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188852
Availability details for
Quantity
  • Availability
  • Add to Cart