ENA429359869 Enamine

1-(1H-1,2,3,4-tetrazol-5-yl)ethan-1-ol

  • MDL number MFCD18810567
  • Cas Number : 155471-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188857

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188857
Availability details for
Quantity
  • Availability
  • Add to Cart