ENA429359870 Enamine

3-(1-benzothiophen-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD14660422
  • Cas Number : 1323441-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188859

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188859
Availability details for
Quantity
  • Availability
  • Add to Cart