ENA429359871 Enamine

5-(1-methylcyclopentyl)-1,2-oxazol-3-amine

  • MDL number MFCD23863461
  • Cas Number : 1501506-59-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188860
Availability details for
Quantity
  • Availability
  • Add to Cart