ENA429359872 Enamine

1-(5-amino-2-methylphenyl)-3-ethylurea

  • MDL number MFCD11213434
  • Cas Number : 1094458-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188861
Availability details for
Quantity
  • Availability
  • Add to Cart