ENA429359876 Enamine

2-(3-amino-4-nitrophenyl)acetic acid

  • MDL number MFCD18389984
  • Cas Number : 1261438-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188867
Availability details for
Quantity
  • Availability
  • Add to Cart