ENA429359877 Enamine

1-[(3-chlorophenyl)methyl]-3-(furan-2-yl)-1H-pyrazol-5-amine

  • MDL number MFCD19127639
  • Cas Number : 1274906-97-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188868

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188868
Availability details for
Quantity
  • Availability
  • Add to Cart