ENA429359878 Enamine

1-(2-hydroxy-4-methylphenyl)propan-1-one

  • MDL number MFCD00218575
  • Cas Number : 2886-52-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188869

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188869
Availability details for
Quantity
  • Availability
  • Add to Cart