ENA429359880 Enamine

3-methyl-3-(4-methylphenyl)pyrrolidine

  • MDL number MFCD16737830
  • Cas Number : 1248449-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188874
Availability details for
Quantity
  • Availability
  • Add to Cart