ENA429359881 Enamine

N-[(2-aminophenyl)methyl]cyclohexanecarboxamide

  • MDL number MFCD15501172
  • Cas Number : 1459704-38-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188876
Availability details for
Quantity
  • Availability
  • Add to Cart