ENA429359882 Enamine

N-(piperidin-4-yl)-1-benzofuran-2-carboxamide hydrochloride

  • MDL number MFCD28118764
  • Cas Number : 150515-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188878
Availability details for
Quantity
  • Availability
  • Add to Cart