ENA429359884 Enamine

2,2,2-trifluoro-1-(oxan-3-yl)ethan-1-ol

  • MDL number MFCD18877729
  • Cas Number : 1342709-61-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188885

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188885
Availability details for
Quantity
  • Availability
  • Add to Cart