ENA429359890 Enamine

1-{5-bromo-1H-pyrrolo[2,3-b]pyridin-3-yl}-2,2,2-trichloroethan-1-one

  • MDL number MFCD22665795
  • Cas Number : 1379670-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188905
Availability details for
Quantity
  • Availability
  • Add to Cart