ENA429359891 Enamine

N-[(2-fluorophenyl)methyl]-1H-pyrazol-4-amine

  • MDL number MFCD12133702
  • Cas Number : 1156177-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-187180
Availability details for
Quantity
  • Availability
  • Add to Cart