ENA429359892 Enamine

2-hydroxy-2-methyl-3-(4-methyl-2-oxo-1,2-dihydropyridin-1-yl)propanoic acid

  • MDL number MFCD20333482
  • Cas Number : 1338977-53-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188915
Availability details for
Quantity
  • Availability
  • Add to Cart