ENA429359894 Enamine

4-fluoro-2-(5-methyl-1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD12800124
  • Cas Number : 1183482-41-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188930

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188930
Availability details for
Quantity
  • Availability
  • Add to Cart