ENA429359895 Enamine

4-(3-methyl-1H-pyrazol-1-yl)piperidine hydrochloride

  • MDL number MFCD28118766
  • Cas Number : 1138820-36-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188931

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188931
Availability details for
Quantity
  • Availability
  • Add to Cart