ENA429359905 Enamine

4-bromo-2-fluoro-3-methylaniline

  • MDL number MFCD26687842
  • Cas Number : 1540204-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188947

Safety Information

msds https://www.enaminestore.com/catalog/EN300-188947
Availability details for
Quantity
  • Availability
  • Add to Cart