ENA429359914 Enamine

1-(1-phenylpiperidin-4-yl)methanamine

  • MDL number MFCD06410624
  • Cas Number : 170353-35-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188956

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188956
Availability details for
Quantity
  • Availability
  • Add to Cart