ENA429359915 Enamine

3-(methoxymethyl)-3-methylazetidine hydrochloride

  • MDL number MFCD28118770
  • Cas Number : 1622351-29-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-188957

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188957
Availability details for
Quantity
  • Availability
  • Add to Cart