ENA429359922 Enamine

lithium(1+) 2-(5-ethoxypyridin-2-yl)acetate

  • MDL number MFCD28118771
  • Cas Number : 1803561-98-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Li+
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-188977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188977
Availability details for
Quantity
  • Availability
  • Add to Cart