ENA429359927 Enamine

2-cyclopropylpropan-1-ol

  • MDL number MFCD21324324
  • Cas Number : 6226-33-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189007

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189007
Availability details for
Quantity
  • Availability
  • Add to Cart