ENA429359932 Enamine

2-(1-hydroxy-1,3-dihydro-2,1-benzoxaborol-3-yl)acetic acid

  • MDL number MFCD20644026
  • Cas Number : 19203-45-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189015
Availability details for
Quantity
  • Availability
  • Add to Cart