ENA429359934 Enamine

2-(aminomethyl)-N-methylpyridin-3-amine

  • MDL number MFCD24131670
  • Cas Number : 1500327-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189019
Availability details for
Quantity
  • Availability
  • Add to Cart