ENA429359935 Enamine

2-[3-(trifluoromethoxy)phenyl]acetaldehyde

  • MDL number MFCD10696558
  • Cas Number : 944898-37-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189020

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189020
Availability details for
Quantity
  • Availability
  • Add to Cart